CAS RN 194-59-2
IUPAC Name:
12-azapentacyclo[11.8.0.0²,¹¹.0³,⁸.0¹⁶,²¹]henicosa-1(13),2(11),3,5,7,9,14,16,18,20-decaene
Synonyms:
7H-dibenzo(C,G)carbazole
Molecular Weight (g/mol):
267.33
Molecular Formula:
C20H13N
InChi Key:
STJXCDGCXVZHDU-UHFFFAOYSA-N
SMILES:
N1C2=C(C3=C1C=CC1=CC=CC=C31)C1=CC=CC=C1C=C2
1
–
2
of
2
results
1
7H-Dibenzo[c,g]carbazole 98.0+%, TCI America™
CAS: 194-59-2 Molecular Formula: C20H13N Molecular Weight (g/mol): 267.331 MDL Number: MFCD00215941 InChI Key: STJXCDGCXVZHDU-UHFFFAOYSA-N Synonym: 7h-dibenzo c,g carbazole,7h-db c,g c,dibenzo c,g carbazole,3,4,5,6-dibenzocarbazole,unii-szf1oji89d,3,4:5,6-dibenzocarbazole,ccris 209,7-aza-7h-dibenzo c,g fluorene PubChem CID: 9134 ChEBI: CHEBI:82312 SMILES: C1=CC=C2C(=C1)C=CC3=C2C4=C(N3)C=CC5=CC=CC=C54
2
7H-Dibenzo(c,g)Carbazole, SPEX CertiPrep™
CAS: 194-59-2 Molecular Formula: C20H13N Molecular Weight (g/mol): 267.331 InChI Key: STJXCDGCXVZHDU-UHFFFAOYSA-N PubChem CID: 9134 ChEBI: CHEBI:82312 SMILES: C1=CC=C2C(=C1)C=CC3=C2C4=C(N3)C=CC5=CC=CC=C54