CAS RN 1937-63-9
CAS RN 1937-63-9
IUPAC Name:
methyl (11E)-octadec-11-enoate
Synonyms:
methyl (11E)-octadec-11-enoate
Molecular Weight (g/mol):
296.5
Molecular Formula:
C19H36O2
InChi Key:
PVVODBCDJBGMJL-CMDGGOBGSA-N
SMILES:
CCCCCC\C=C\CCCCCCCCCC(=O)OC
1
–
3
of
3
results
2
Methyl cis-11-octadecenoate, ≥99% (GC), MilliporeSigma™ Supelco™
Methyl cis-11-octadecenoate is a fatty acid methyl ester.
3
Methyl cis-11-Octadecenoate 98.0+%, TCI America™
CAS: 1937-63-9 Molecular Formula: C19H36O2 Molecular Weight (g/mol): 296.50 MDL Number: MFCD00010457,MFCD00064484 InChI Key: PVVODBCDJBGMJL-CMDGGOBGSA-N Synonym: methyl cis-11-octadecenoate,cis-11-octadecenoic acid methyl ester,methyl z-octadec-11-enoate,methyl cis-vaccenate,cis-vaccenic acid methyl ester,11-octadecenoic acid, methyl ester, z,methyl 11z-octadec-11-enoate,methyl octadec-11-enoate,methyl asclepate,methyl cis-octadec-11-enoate PubChem CID: 5364505 IUPAC Name: methyl (11E)-octadec-11-enoate SMILES: CCCCCC\C=C\CCCCCCCCCC(=O)OC