CAS RN 19353-92-5
IUPAC Name:
4-(dimethylamino)benzohydrazide
Synonyms:
4-(dimethylamino)benzohydrazide
Molecular Weight (g/mol):
179.22
Molecular Formula:
C9H13N3O
InChi Key:
HITIGLAGJBMISF-UHFFFAOYSA-N
SMILES:
CN(C)C1=CC=C(C=C1)C(=O)NN
1
–
1
of
1
results
1
Promotions Available
4-Dimethylaminobenzhydrazide, 98+%
4-Dimethylaminobenzhydrazide, 98+%
CAS: 19353-92-5 Molecular Formula: C9H13N3O Molecular Weight (g/mol): 179.223 MDL Number: MFCD00014761 InChI Key: HITIGLAGJBMISF-UHFFFAOYSA-N Synonym: 4-dimethylamino benzohydrazide,4-dimethylamino benzhydrazide,4-dimethylaminobenzhydrazide,p-dimethylamino benzohydrazide,4-n,n-dimethylamino benzhydrazide,dimethyl 4-???phenyl amine,4me2nphcon2,acmc-1bqxb,maybridge1_002636,4-dimethylaminobenzoic acid, hydrazide PubChem CID: 88019 IUPAC Name: 4-(dimethylamino)benzohydrazide SMILES: CN(C)C1=CC=C(C=C1)C(=O)NN