missing translation for 'onlineSavingsMsg'
Learn More

4-Dimethylaminobenzhydrazide, 98+%

Catalog No. AAA1372318
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1372318 50 g
AAA1372309 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1372318 Supplier Thermo Scientific Chemicals Supplier No. A1372318
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 19353-92-5 | C9H13N3O | 179.223 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19353-92-5
Molecular Formula C9H13N3O
Molecular Weight (g/mol) 179.223
MDL Number MFCD00014761
InChI Key HITIGLAGJBMISF-UHFFFAOYSA-N
Synonym 4-dimethylamino benzohydrazide, 4-dimethylamino benzhydrazide, 4-dimethylaminobenzhydrazide, p-dimethylamino benzohydrazide, 4-n,n-dimethylamino benzhydrazide, dimethyl 4-???phenyl amine, 4me2nphcon2, acmc-1bqxb, maybridge1_002636, 4-dimethylaminobenzoic acid, hydrazide
PubChem CID 88019
IUPAC Name 4-(dimethylamino)benzohydrazide
SMILES CN(C)C1=CC=C(C=C1)C(=O)NN

Specifications

Melting Point 174°C to 175°C
Quantity 50 g
Beilstein 2210856
Formula Weight 179.22
Percent Purity ≥98%
Chemical Name or Material 4-Dimethylaminobenzhydrazide
Hazard Category H302-H317-H351
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P272-P280g-P281-P301+P312-P302+P352-P304+P340-P308+P313-P330-P333+P313-P363-P501c
EINECSNumber 242-985-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only