accessibility menu, dialog, popup

UserName

CAS RN 192998-62-2

LiFFFSOONSOOF

CAS RN 192998-62-2

IUPAC Name: lithium(1+) (fluorosulfonyl)(trifluoromethanesulfonyl)azanide
Synonyms: lithium(1+) (fluorosulfonyl)(trifluoromethanesulfonyl)azanide
Molecular Weight (g/mol): 237.07
Molecular Formula: CF4LiNO4S2
InChi Key: ZIRAMZRKLHPLPK-UHFFFAOYSA-N
SMILES: [Li+].FC(F)(F)S(=O)(=O)[N-]S(F)(=O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results