missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium (Fluorosulfonyl)(trifluoromethanesulfonyl)imide 95.0+%, TCI America™
Supplier: TCI America L02951G
Specifications
| Lithium (Fluorosulfonyl)(trifluoromethanesulfonyl)imide | |
| 100°C | |
| CF4LiNO4S2 | |
| 1759 | |
| [Li+].C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)F | |
| 237.068 | |
| 237.07 | |
| Crystalline Powder |
| 192998-62-2 | |
| White | |
| 1 g | |
| ZIRAMZRKLHPLPK-UHFFFAOYSA-N | |
| lithium;fluorosulfonyl(trifluoromethylsulfonyl)azanide | |
| 101539524 | |
| ≥97.0% (T) |
Chemical Identifiers
| 192998-62-2 | |
| 237.068 | |
| 101539524 | |
| [Li+].C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)F |
| CF4LiNO4S2 | |
| ZIRAMZRKLHPLPK-UHFFFAOYSA-N | |
| lithium;fluorosulfonyl(trifluoromethylsulfonyl)azanide |
Safety and Handling
TSCA : No