accessibility menu, dialog, popup

UserName

CAS RN 1929-29-9

IUPAC Name: 3-(4-methoxyphenyl)propanoic acid
Synonyms: 2-(p-anisyl)acetic acid
Molecular Weight (g/mol): 180.2
Molecular Formula: C10H12O3
InChi Key: FIUFLISGGHNPSM-UHFFFAOYSA-N
SMILES: COC1=CC=C(CCC(O)=O)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)