missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Methoxyphenyl)propionic acid, 98%

Catalog No. AAA1365522
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1365522 100 g
AAA1365514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1365522 Supplier Thermo Scientific Chemicals Supplier No. A1365522
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1929-29-9 | C10H12O3 | 180.203 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1929-29-9
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00002777
InChI Key FIUFLISGGHNPSM-UHFFFAOYSA-N
Synonym 3-4-methoxyphenyl propanoic acid, 3-4-methoxyphenyl propionic acid, benzenepropanoic acid, 4-methoxy, p-methoxyhydrocinnamic acid, 3-p-methoxyphenyl propionic acid, 3-4-methoxy-phenyl-propionic acid, 4-methoxyhydrocinnamic acid, 3-4-methoxyphenyl-propionic acid, 4-methoxyhydro cinnamic acid, hydrocinnamic acid, p-methoxy
PubChem CID 95750
IUPAC Name 3-(4-methoxyphenyl)propanoic acid
SMILES COC1=CC=C(C=C1)CCC(=O)O

Specifications

Melting Point 102°C to 106°C
Quantity 100 g
Beilstein 1953326
Formula Weight 180.2
Percent Purity 98%
Chemical Name or Material 3-(4-Methoxyphenyl)propionic acid
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 217-678-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only