accessibility menu, dialog, popup

UserName

CAS RN 19284-81-2

NH2SNOO

CAS RN 19284-81-2

IUPAC Name: 2-[(2-nitrophenyl)sulfanyl]aniline
Synonyms: 2-[(2-nitrophenyl)sulfanyl]aniline
Molecular Weight (g/mol): 246.28
Molecular Formula: C12H10N2O2S
InChi Key: QREMWHOIUTWWSC-UHFFFAOYSA-N
SMILES: NC1=CC=CC=C1SC1=CC=CC=C1[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results