missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-2'-nitrodiphenyl Sulfide 98.0+%, TCI America™

Catalog No. A19855G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A19855G 5 g
A198525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A19855G Supplier TCI America Supplier No. A19855G
Only null left

Chemical Identifiers

CAS 19284-81-2
Molecular Formula C12H10N2O2S
Molecular Weight (g/mol) 246.28
MDL Number MFCD00121816
InChI Key QREMWHOIUTWWSC-UHFFFAOYSA-N
Synonym 2-(2-Nitrophenylthio)aniline, 2-Aminophenyl 2-Nitrophenyl Sulfide
PubChem CID 699904
IUPAC Name 2-[(2-nitrophenyl)sulfanyl]aniline
SMILES NC1=CC=CC=C1SC1=CC=CC=C1[N+]([O-])=O

Specifications

Melting Point 87°C
Color Yellow
Quantity 5 g
Formula Weight 246.28
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-2′-nitrodiphenyl Sulfide