accessibility menu, dialog, popup

UserName

CAS RN 19220-93-0

CH3OOFFFFF

CAS RN 19220-93-0

IUPAC Name: 2,3,4,5,6-pentafluorophenyl acetate
Synonyms: 2,3,4,5,6-pentafluorophenyl acetate
Molecular Weight (g/mol): 226.1
Molecular Formula: C8H3F5O2
InChi Key: ZXTVBLZVILLKPM-UHFFFAOYSA-N
SMILES: CC(=O)OC1=C(F)C(F)=C(F)C(F)=C1F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results