missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorophenyl Acetate 98.0+%, TCI America™

Catalog No. P21925G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P21925G 5 g
P21921G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P21925G Supplier TCI America Supplier No. P21925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 19220-93-0
Molecular Formula C8H3F5O2
Molecular Weight (g/mol) 226.10
MDL Number MFCD00015552
InChI Key ZXTVBLZVILLKPM-UHFFFAOYSA-N
Synonym Acetic Acid Pentafluorophenyl Ester
PubChem CID 87968
IUPAC Name 2,3,4,5,6-pentafluorophenyl acetate
SMILES CC(=O)OC1=C(F)C(F)=C(F)C(F)=C1F

Specifications

Melting Point 32°C
Color White
Boiling Point 60°C
Quantity 5 g
Formula Weight 226.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Pentafluorophenyl Acetate