accessibility menu, dialog, popup

UserName

CAS RN 19094-56-5

IUPAC Name: 2-chloro-5-iodobenzoic acid
Synonyms: 2-chloro-5-iodobenzoic acid
Molecular Weight (g/mol): 282.46
Molecular Formula: C7H4ClIO2
InChi Key: GEBYSTBEDVQOTK-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(Cl)C=CC(I)=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)