missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-iodobenzoic Acid 97.0+%, TCI America™

Catalog No. C21685G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C21685G 5 g
C216825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C21685G Supplier TCI America Supplier No. C21685G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 19094-56-5
Molecular Formula C7H4ClIO2
Molecular Weight (g/mol) 282.46
MDL Number MFCD00079731
InChI Key GEBYSTBEDVQOTK-UHFFFAOYSA-N
Synonym 2-chloro-5-iodo-benzoic acid, benzoic acid, 2-chloro-5-iodo, pubchem3649, acmc-209eug, 5-iodo-2-chlorobenzoic acid, ksc174i1h, benzoicacid, 2-chloro-5-iodo, buttpark 100\01-43, 2-chloro-5-iodobenzoic acid
PubChem CID 519638
IUPAC Name 2-chloro-5-iodobenzoic acid
SMILES OC(=O)C1=C(Cl)C=CC(I)=C1

Specifications

Melting Point 160°C
Color White-Yellow
Quantity 5 g
Formula Weight 282.46
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-5-iodobenzoic Acid