CAS RN 19049-40-2
IUPAC Name:
tetraberyllium(2+) hexaacetate oxidandiide
Synonyms:
basic beryllium acetate
Molecular Weight (g/mol):
406.31
Molecular Formula:
C12H18Be4O13
InChi Key:
RKARILOACIIVIJ-UHFFFAOYSA-H
SMILES:
[Be++].[Be++].[Be++].[Be++].[O--].CC([O-])=O.CC([O-])=O.CC([O-])=O.CC([O-])=O.CC([O-])=O.CC([O-])=O
1
–
3
of
3
results
1
Beryllium, AAS standard solution, Specpure™ Be 1000μg/mL
CAS: 19049-40-2 Molecular Formula: Be in 5% HNO3 MDL Number: MFCD00134032
2
Beryllium ICP Standard, 1000 ppm Be in 3% HNO3, Ricca Chemical
CAS: 19049-40-2
3
Beryllium ICP Standard, 10,000 ppm Be in 5% HNO3, Ricca Chemical
CAS: 19049-40-2