missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beryllium ICP Standard, 10,000 ppm Be in 5% HNO3, Ricca Chemical
| Catalog Number | Mfr. No. | dimQuantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | dimQuantity | Packaging | Price | Quantity & Availability | ||||
|
PBE10KN50
|
N/A | Natural Poly Bottle |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
PBE10KN100
|
N/A | Natural LDPE Bottle |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 19049-40-2, 7697-37-2, 19049-40-2 |
Specifications
| 19049-40-2 , 7697-37-2 , 19049-40-2 | |
| 70.0, 20.01, 7.9 | |
| 1 mL = 10 mg Be |
| 10,000ppm Be | |
| 73.0, 20.83, 8.23 | |
| Beryllium ICP Standard |