accessibility menu, dialog, popup

UserName

CH3NH2NN

CAS RN 1904-31-0

IUPAC Name: 1-methyl-1H-pyrazol-3-amine
Synonyms: 1-methylpyrazol-3-amine
Molecular Weight (g/mol): 97.12
Molecular Formula: C4H7N3
InChi Key: MOGQNVSKBCVIPW-UHFFFAOYSA-N
SMILES: CN1C=CC(N)=N1
Boiling Point 
  • (2)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (1)
  • (3)
  • (2)