accessibility menu, dialog, popup

UserName

CH3OOOCH3

CAS RN 18927-05-4

IUPAC Name: methyl 2-(3-methoxyphenyl)acetate
Synonyms: methyl 2-(3-methoxyphenyl)acetate
Molecular Weight (g/mol): 180.2
Molecular Formula: C10H12O3
InChi Key: BSVIOYCZTJRBDB-UHFFFAOYSA-N
SMILES: COC(=O)CC1=CC=CC(OC)=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)