missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-Methoxyphenylacetate 98.0+%, TCI America™

Catalog No. M13315G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M13315G 5 g
M133125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M13315G Supplier TCI America Supplier No. M13315G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 18927-05-4
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00017205
InChI Key BSVIOYCZTJRBDB-UHFFFAOYSA-N
Synonym methyl 2-3-methoxyphenyl acetate, methyl 3-methoxyphenylacetate, benzeneacetic acid, 3-methoxy-, methyl ester, acetic acid, m-methoxyphenyl-, methyl ester, methyl 3-methoxyphenyl acetate, 3-methoxy-phenyl-acetic acid methyl ester, acmc-1brer, methyl m-methoxyphenylacetate, methyl 3-methoxy-phenylacetate
PubChem CID 519609
IUPAC Name methyl 2-(3-methoxyphenyl)acetate
SMILES COC1=CC=CC(=C1)CC(=O)OC

Specifications

Color Yellow
Quantity 5 g
Formula Weight 180.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 3-Methoxyphenylacetate