accessibility menu, dialog, popup

UserName

RbFBFFF

CAS RN 18909-68-7

IUPAC Name: rubidium(1+) tetrafluoroboranuide
Synonyms: rubidium(1+) tetrafluoroborate
Molecular Weight (g/mol): 172.27
Molecular Formula: BF4Rb
InChi Key: MERZTPVDEIGCOQ-UHFFFAOYSA-N
SMILES: [Rb+].F[B-](F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results