missing translation for 'onlineSavingsMsg'
Learn More

Rubidium tetrafluoroborate, 98%

Catalog No. AAB2081714
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2081714 25 g
AAB2081706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2081714 Supplier Thermo Scientific Chemicals Supplier No. B2081714

CAS: 18909-68-7 | BF4Rb | 172.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 18909-68-7
Molecular Formula BF4Rb
Molecular Weight (g/mol) 172.27
MDL Number MFCD00083071
InChI Key MERZTPVDEIGCOQ-UHFFFAOYSA-N
Synonym rubidium tetrafluoroborate, rubidium 1+ tetrafluoroborate, rubidium fluoroborate, rubidium boron fluoride, borate 1-, rubidium, acmc-1bu19, rubidium 1+ ion tetrafluoroborate
PubChem CID 23696894
IUPAC Name rubidium(1+) tetrafluoroboranuide
SMILES [Rb+].F[B-](F)(F)F

Specifications

Quantity 25 g
Linear Formula RbBF4
Formula Weight 172.27
Percent Purity 98%
Chemical Name or Material Rubidium tetrafluoroborate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 242-660-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only