accessibility menu, dialog, popup

UserName

H3COBr

CAS RN 1889-26-5

IUPAC Name: pentanoyl bromide
Synonyms: pentanoyl bromide
Molecular Weight (g/mol): 165.03
Molecular Formula: C5H9BrO
InChi Key: FDOHPUYPQQKECS-UHFFFAOYSA-N
SMILES: CCCCC(Br)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results