missing translation for 'onlineSavingsMsg'
Learn More

Valeryl Bromide 98.0+%, TCI America™

Catalog No. V003810G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V003810G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. V003810G Supplier TCI America Supplier No. V003810G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1889-26-5
Molecular Formula C5H9BrO
Molecular Weight (g/mol) 165.03
MDL Number MFCD00059467
InChI Key FDOHPUYPQQKECS-UHFFFAOYSA-N
Synonym Valeroyl Bromide
PubChem CID 11137484
IUPAC Name pentanoyl bromide
SMILES CCCCC(=O)Br

Specifications

Boiling Point 64°C
Quantity 10 g
UN Number 2920
Formula Weight 165.03
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Valeryl Bromide