accessibility menu, dialog, popup

UserName

CAS RN 188011-69-0

IUPAC Name: 3-[(5-bromopyridin-2-yl)carbamoyl]propanoic acid
Synonyms: 3-[(5-bromopyridin-2-yl)carbamoyl]propanoic acid
Molecular Weight (g/mol): 273.09
Molecular Formula: C9H9BrN2O3
InChi Key: XFYYQDHEDOXWGA-UHFFFAOYSA-N
SMILES: OC(=O)CCC(=O)NC1=NC=C(Br)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results