missing translation for 'onlineSavingsMsg'
Learn More

Bikinin 98.0+%, TCI America™

Catalog No. B482625MG
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B482625MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B482625MG Supplier TCI America Supplier No. B482625MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 188011-69-0
Molecular Formula C9H9BrN2O3
Molecular Weight (g/mol) 273.086
MDL Number MFCD00116442
InChI Key XFYYQDHEDOXWGA-UHFFFAOYSA-N
Synonym 4-[(5-Bromo-2-pyridyl)amino]-4-oxobutyric Acid
PubChem CID 647833
IUPAC Name 4-[(5-bromopyridin-2-yl)amino]-4-oxobutanoic acid
SMILES C1=CC(=NC=C1Br)NC(=O)CCC(=O)O

Specifications

Melting Point 144°C
Color White
Quantity 25 mg
Formula Weight 273.09
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Bikinin
TSCA No
Recommended Storage Refrigerator