CAS RN 187947-37-1
IUPAC Name:
N-[2-(5-hydroxy-1H-indol-3-yl)ethyl]icosa-5,8,11,14-tetraenamide
Synonyms:
arachidonoyl serotonin
Molecular Weight (g/mol):
462.68
Molecular Formula:
C30H42N2O2
InChi Key:
QJDNHGXNNRLIGA-UHFFFAOYSA-N
SMILES:
CCCCCC=CCC=CCC=CCC=CCCCC(=O)NCCC1=CNC2=CC=C(O)C=C12
1
–
1
of
1
results
1
N-Arachidonoyl-serotonin, 98%, Thermo Scientific Chemicals
CAS: 187947-37-1 Molecular Formula: C30H42N2O2 Molecular Weight (g/mol): 462.68 MDL Number: MFCD02179189 InChI Key: QJDNHGXNNRLIGA-UHFFFAOYSA-N Synonym: n-arachidonoyl-serotonin PubChem CID: 53394325 IUPAC Name: N-[2-(5-hydroxy-1H-indol-3-yl)ethyl]icosa-5,8,11,14-tetraenamide SMILES: CCCCCC=CCC=CCC=CCC=CCCCC(=O)NCCC1=CNC2=CC=C(O)C=C12