accessibility menu, dialog, popup

UserName

OHONOO

CAS RN 1877-73-2

IUPAC Name: 2-(3-nitrophenyl)acetic acid
Synonyms: 2-(3-nitrophenyl)acetic acid M-nitrophenylacetic acid
Molecular Weight (g/mol): 181.15
Molecular Formula: C8H7NO4
InChi Key: WUKHOVCMWXMOOA-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC=CC(=C1)[N+]([O-])=O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)