missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrophenylacetic acid, 99%

Catalog No. AAB2154014
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2154014 25 g
AAB2154003 1 g
AAB2154006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2154014 Supplier Thermo Scientific Chemicals Supplier No. B2154014
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1877-73-2 | C8H7NO4 | 181.147 g/mol

3-Nitrophenylacetic acid, is a useful synthetic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Nitrophenylacetic acid, is a useful synthetic intermediate.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong bases.

Chemical Identifiers

CAS 1877-73-2
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.147
MDL Number MFCD00007278
InChI Key WUKHOVCMWXMOOA-UHFFFAOYSA-N
Synonym 3-nitrophenylacetic acid, 2-3-nitrophenyl acetic acid, 3-nitrophenyl acetic acid, benzeneacetic acid, 3-nitro, m-nitrophenyl acetic acid, m-nitrophenylacetic acid, 3-nitrobenzeneacetic acid, acetic acid, m-nitrophenyl, ccris 2337
PubChem CID 15876
SMILES C1=CC(=CC(=C1)[N+](=O)[O-])CC(=O)O

Specifications

Melting Point 117°C to 120°C
Quantity 25 g
Beilstein 2050088
Solubility Information Soluble in water.
Formula Weight 181.15
Percent Purity 99%
Chemical Name or Material 3-Nitrophenylacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 217-512-7
RTECSNumber AJ1129900
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only