accessibility menu, dialog, popup

UserName

CH3OOOHO

CAS RN 1877-71-0

IUPAC Name: 3-(methoxycarbonyl)benzoic acid
Synonyms: isophthalic acid, methyl ester
Molecular Weight (g/mol): 180.16
Molecular Formula: C9H8O4
InChi Key: WMZNGTSLFSJHMZ-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC(=C1)C(O)=O
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (4)
  • (4)
  • (2)
  • (2)
  • (4)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)