missing translation for 'onlineSavingsMsg'
Learn More

Methyl hydrogen isophthalate, 97%

Catalog No. AAL1299206
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1299206 5 g
AAL1299203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1299206 Supplier Thermo Scientific Chemicals Supplier No. L1299206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1877-71-0 | C9H8O4 | 180.159 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1877-71-0
Molecular Formula C9H8O4
Molecular Weight (g/mol) 180.159
MDL Number MFCD00029972
InChI Key WMZNGTSLFSJHMZ-UHFFFAOYSA-N
Synonym 3-methoxycarbonyl benzoic acid, mono-methyl isophthalate, methyl hydrogen isophthalate, isophthalic acid monomethyl ester, monomethyl isophthalate, isophthalic acid methyl ester, isophthalic acid, methyl ester, isophthalic acid, monomethyl ester, methyl m-phthalate, methyl 3-carboxybenzoate
PubChem CID 601880
IUPAC Name 3-methoxycarbonylbenzoic acid
SMILES COC(=O)C1=CC=CC(=C1)C(=O)O

Specifications

Melting Point 193°C to 194°C
Quantity 5 g
Beilstein 2048750
Formula Weight 180.16
Percent Purity 97%
Chemical Name or Material Methyl hydrogen isophthalate
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only