accessibility menu, dialog, popup

UserName

BrBr

CAS RN 186517-01-1

IUPAC Name: 2,6-dibromoanthracene
Synonyms: 2,6-dibromoanthracene
Molecular Weight (g/mol): 336.03
Molecular Formula: C14H8Br2
InChi Key: BPRGLVVFWRNXEP-UHFFFAOYSA-N
SMILES: BrC1=CC2=CC3=CC=C(Br)C=C3C=C2C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results