missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dibromoanthracene 98.0+%, TCI America™
Specifications
Specifications
| Quantity | 5 g |
| Formula Weight | 336.03 |
| Percent Purity | ≥98.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2,6-Dibromoanthracene |
Safety and Handling
| TSCA | No |