accessibility menu, dialog, popup

UserName

CH3H3CSN

CAS RN 18640-74-9

IUPAC Name: 2-(2-methylpropyl)-1,3-thiazole
Synonyms: isobutylthiazole
Molecular Weight (g/mol): 141.23
Molecular Formula: C7H11NS
InChi Key: CMPVUVUNJQERIT-UHFFFAOYSA-N
SMILES: CC(C)CC1=NC=CS1
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)