accessibility menu, dialog, popup

UserName

CAS RN 18595-12-5

IUPAC Name: methyl 5-amino-2-methylbenzoate
Synonyms: methyl 5-amino-2-methylbenzoate
Molecular Weight (g/mol): 165.19
Molecular Formula: C9H11NO2
InChi Key: JNPZKGOLYSCSEL-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(C)C=CC(N)=C1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)