missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-amino-2-methylbenzoate, 97%

Catalog No. AAH6167122
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6167122 100 g
AAH6167114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6167122 Supplier Thermo Scientific Chemicals Supplier No. H6167122
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 18595-12-5 | C9H11NO2 | 165.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 18595-12-5
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
MDL Number MFCD08752568
InChI Key JNPZKGOLYSCSEL-UHFFFAOYSA-N
PubChem CID 15049977
IUPAC Name methyl 5-amino-2-methylbenzoate
SMILES COC(=O)C1=C(C)C=CC(N)=C1

Specifications

Melting Point 39°C to 40°C
Color Brown to Red
Quantity 100 g
Beilstein 2716876
Synonym 5-amino-2-methyl-benzoic acid methyl ester, 5-amino-2-methylbenzoic acid methyl ester, 5-amino-2-methyl benzoic acid methyl ester, benzoic acid,5-amino-2-methyl-, methyl ester, benzoic acid, 5-amino-2-methyl-, methyl ester, 2-methyl-5-aminobenzoicacidmethylester, methyl 5-amino-o-toluate, methyl 2-methyl-5-aminobenzoate, methyl 3-amino-6-methylbenzoate, methyl 5-amino-2-methyl-benzoate
Formula Weight 165.19
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Methyl 5-amino-2-methylbenzoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only