accessibility menu, dialog, popup

UserName

CAS RN 18529-04-9

H3CNFCl

CAS RN 18529-04-9

IUPAC Name: 4-chloro-7-fluoro-2-methylquinoline
Synonyms: 4-chloro-7-fluoro-2-methylquinoline
Molecular Weight (g/mol): 195.62
Molecular Formula: C10H7ClFN
InChi Key: BBZQRRHOPBBNRO-UHFFFAOYSA-N
SMILES: CC1=CC(Cl)=C2C=CC(F)=CC2=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results