accessibility menu, dialog, popup

UserName

BrOBr

CAS RN 18523-22-3

IUPAC Name: 2-bromo-1-(3-bromophenyl)ethan-1-one
Synonyms: 2-bromo-1-(3-bromophenyl)ethanone
Molecular Weight (g/mol): 277.94
Molecular Formula: C8H6Br2O
InChi Key: MZBXSQBQPJWECM-UHFFFAOYSA-N
SMILES: BrCC(=O)C1=CC=CC(Br)=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)