CAS RN 18306-29-1
CAS RN 18306-29-1
IUPAC Name:
ditrimethylsilyl sulfate
Synonyms:
ditrimethylsilyl sulfate
Molecular Weight (g/mol):
242.44
Molecular Formula:
C6H18O4SSi2
InChi Key:
KRUQDZRWZXUUAD-UHFFFAOYSA-N
SMILES:
C[Si](C)(C)OS(=O)(=O)O[Si](C)(C)C
1
–
1
of
1
results
1
Bis(trimethylsilyl) Sulfate 98.0+%, TCI America™
CAS: 18306-29-1 Molecular Formula: C6H18O4SSi2 Molecular Weight (g/mol): 242.436 MDL Number: MFCD00008268 InChI Key: KRUQDZRWZXUUAD-UHFFFAOYSA-N Synonym: bis trimethylsilyl sulfate,bistrimethylsilylsulfate,ditrimethylsilyl sulfate,sulfuric acid, tms,bis trimethylsilyl sulphate,silanol, trimethyl-, sulfate 2:1,acmc-209ekb,bis-trimethylsilyl-sulfate PubChem CID: 87573 IUPAC Name: bis(trimethylsilyl) sulfate SMILES: C[Si](C)(C)OS(=O)(=O)O[Si](C)(C)C