accessibility menu, dialog, popup

UserName

CAS RN 1820-99-1

HOOSOHHOOHO

CAS RN 1820-99-1

IUPAC Name: 5-[(3-carboxy-4-hydroxyphenyl)sulfanyl]-2-hydroxybenzoic acid
Synonyms: 5-[(3-carboxy-4-hydroxyphenyl)sulfanyl]-2-hydroxybenzoic acid
Molecular Weight (g/mol): 306.29
Molecular Formula: C14H10O6S
InChi Key: DWMJRSPNFCPIQN-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(SC2=CC=C(O)C(=C2)C(O)=O)=CC=C1O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results