missing translation for 'onlineSavingsMsg'
Learn More

5,5'-Thiodisalicylic Acid 98.0+%, TCI America™

Catalog No. T02085G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T02085G 5 g
T020825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T02085G Supplier TCI America Supplier No. T02085G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1820-99-1
Molecular Formula C14H10O6S
Molecular Weight (g/mol) 306.288
MDL Number MFCD00020264
InChI Key DWMJRSPNFCPIQN-UHFFFAOYSA-N
PubChem CID 74562
IUPAC Name 5-(3-carboxy-4-hydroxyphenyl)sulfanyl-2-hydroxybenzoic acid
SMILES C1=CC(=C(C=C1SC2=CC(=C(C=C2)O)C(=O)O)C(=O)O)O

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 306.29
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5,5′-Thiodisalicylic Acid