accessibility menu, dialog, popup

UserName

CAS RN 181705-93-1

CFZnBr

CAS RN 181705-93-1

IUPAC Name: 4-fluorobenzen-1-ide; bromozincylium
Synonyms: 4-fluorobenzen-1-ide; bromozincylium
Molecular Weight (g/mol): 240.38
Molecular Formula: C6H4BrFZn
InChi Key: VRJVWWPYGMESMA-UHFFFAOYSA-M
SMILES: [Zn+]Br.FC1=CC=[C-]C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results