missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluorophenylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Density | 0.992 |
| Quantity | 50 mL |
| UN Number | UN3399 |
| Sensitivity | Air sensitive |
| Formula Weight | 240.38 |
| Chemical Name or Material | 4-Fluorophenylzinc bromide, Packaged under Argon in resealable ChemSeal™ bottles |
Safety and Handling
| Hazard Category | H225-H260-H302-H314-H335-H336-H351 |
| Hazard Statement | GHS H Statement H225-H261-H314-H351-H335 Highly flammable liquid and vapor. In contact with water releases flammable gas. Causes severe skin burns and eye damage. Suspected of causing cancer. May cause respiratory irritation. |
| Precautionary Statement | P201-P202-P210-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE |
| TSCA | No |
| Recommended Storage | Keep cold; Store under Argon |
RUO – Research Use Only