accessibility menu, dialog, popup

UserName

H3CSNNSCH3

CAS RN 180729-91-3

IUPAC Name: 4-nonyl-2-(4-nonyl-1,3-thiazol-2-yl)-1,3-thiazole
Synonyms: 4-nonyl-2-(4-nonyl-1,3-thiazol-2-yl)-1,3-thiazole
Molecular Weight (g/mol): 420.72
Molecular Formula: C24H40N2S2
InChi Key: FNAZWICXOXNWDC-UHFFFAOYSA-N
SMILES: CCCCCCCCCC1=CSC(=N1)C1=NC(CCCCCCCCC)=CS1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results