missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dinonyl-2,2'-bithiazole 98.0+%, TCI America™

Catalog No. D4559200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4559200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D4559200MG Supplier TCI America Supplier No. D4559200MG
Only null left

Chemical Identifiers

CAS 180729-91-3
Molecular Formula C24H40N2S2
Molecular Weight (g/mol) 420.72
MDL Number MFCD27923057
InChI Key FNAZWICXOXNWDC-UHFFFAOYSA-N
PubChem CID 19437291
IUPAC Name 4-nonyl-2-(4-nonyl-1,3-thiazol-2-yl)-1,3-thiazole
SMILES CCCCCCCCCC1=CSC(=N1)C1=NC(CCCCCCCCC)=CS1

Specifications

Melting Point 61°C
Color Yellow
Quantity 200 mg
Formula Weight 420.72
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dinonyl-2,2′-bithiazole