accessibility menu, dialog, popup

UserName

CAS RN 180675-22-3

IUPAC Name: 2-[(bromomagnesio)methyl]-1,3-dioxolane
Synonyms: 2-[(bromomagnesio)methyl]-1,3-dioxolane
Molecular Weight (g/mol): 191.31
Molecular Formula: C4H7BrMgO2
InChi Key: ZDNTZRAFJZERSL-UHFFFAOYSA-M
SMILES: Br[Mg]CC1OCCO1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)