accessibility menu, dialog, popup

UserName

NONO

CAS RN 180186-94-1

IUPAC Name: 2-({3aH,8H,8aH-indeno[1,2-d][1,3]oxazol-2-yl}methyl)-3aH,8H,8aH-indeno[1,2-d][1,3]oxazole
Synonyms: 2-{3aH,8H,8aH-indeno[1,2-d][1,3]oxazol-2-ylmethyl}-3aH,8H,8aH-indeno[1,2-d][1,3]oxazole
Molecular Weight (g/mol): 330.39
Molecular Formula: C21H18N2O2
InChi Key: BDHSVQLSNIGJNC-UHFFFAOYNA-N
SMILES: C(C1=NC2C(CC3=CC=CC=C23)O1)C1=NC2C(CC3=CC=CC=C23)O1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)