missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(+)-2,2'-Methylenebis[(3aR,8aS)-3a,8a-dihydro-8H-indeno[1,2-d]oxazole] 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 225°C |
| Color | White-Yellow |
| Quantity | 500 mg |
| Formula Weight | 330.39 |
| Percent Purity | ≥98.0% (HPLC,N) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | (+)-2,2′-Methylenebis[(3aR,8aS)-3a,8a-dihydro-8H-indeno[1,2-d]oxazole] |
Safety and Handling
| TSCA | No |