accessibility menu, dialog, popup

UserName

CAS RN 17978-77-7

CH3H3CCH3OFFFFFFFOHCH3H3CCH3OFFFFFFFOHCH3H3CCH3OFFFFFFFOHPr(E/Z)(E/Z)(E/Z)

CAS RN 17978-77-7

IUPAC Name: tris(1,1,1,2,2,3,3-heptafluoro-6-hydroxy-7,7-dimethyloct-5-en-4-one) praseodymium
Synonyms: tris(1,1,1,2,2,3,3-heptafluoro-6-hydroxy-7,7-dimethyloct-5-en-4-one) praseodymium
Molecular Weight (g/mol): 1029.46
Molecular Formula: C30H33F21O6Pr
InChi Key: LICWBBABBAOMPH-UHFFFAOYSA-N
SMILES: [Pr].CC(C)(C)C(O)=CC(=O)C(F)(F)C(F)(F)C(F)(F)F.CC(C)(C)C(O)=CC(=O)C(F)(F)C(F)(F)C(F)(F)F.CC(C)(C)C(O)=CC(=O)C(F)(F)C(F)(F)C(F)(F)F
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results