missing translation for 'onlineSavingsMsg'
Learn More

Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato)praseodymium(III) 97.0+%, TCI America™

Catalog No. T15111G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T15111G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T15111G Supplier TCI America Supplier No. T15111G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 17978-77-7
Molecular Formula C30H33F21O6Pr
Molecular Weight (g/mol) 1029.462
MDL Number MFCD00064656
InChI Key LICWBBABBAOMPH-VNGPFPIXSA-N
Synonym praseodymium 6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionate
PubChem CID 131668219
IUPAC Name (Z)-1,1,1,2,2,3,3-heptafluoro-6-hydroxy-7,7-dimethyloct-5-en-4-one;praseodymium
SMILES CC(C)(C)C(=CC(=O)C(C(C(F)(F)F)(F)F)(F)F)O.CC(C)(C)C(=CC(=O)C(C(C(F)(F)F)(F)F)(F)F)O.CC(C)(C)C(=CC(=O)C(C(C(F)(F)F)(F)F)(F)F)O.[Pr]

Specifications

Melting Point 219°C
Color Green-Yellow
Quantity 1 g
Formula Weight 1026.44
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato)praseodymium(III) [NMR Shift Reagent]