accessibility menu, dialog, popup

UserName

H3CSCH3O

CAS RN 1778-09-2

IUPAC Name: 1-[4-(methylsulfanyl)phenyl]ethan-1-one
Synonyms: 1-[4-(methylsulfanyl)phenyl]ethanone
Molecular Weight (g/mol): 166.24
Molecular Formula: C9H10OS
InChi Key: JECUZQLBQKNEMW-UHFFFAOYSA-N
SMILES: CSC1=CC=C(C=C1)C(C)=O
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)